Skip to Content
Merck
All Photos(1)

Key Documents

48543

Supelco

Hexachloro-1,3-butadiene

analytical standard

Synonym(s):

Perchlorobutadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CCl2=CClCCl=CCl2
CAS Number:
Molecular Weight:
260.76
Beilstein:
1766570
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

0.2 mmHg ( 20 °C)

CofA

current certificate can be downloaded

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.555 (lit.)

bp

210-220 °C (lit.)

mp

−22-−19 °C (lit.)

density

1.665 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Cl\C(Cl)=C(Cl)/C(Cl)=C(\Cl)Cl

InChI

1S/C4Cl6/c5-1(3(7)8)2(6)4(9)10

InChI key

RWNKSTSCBHKHTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B Staples et al.
Occupational and environmental medicine, 60(7), 463-467 (2003-06-24)
To determine the prevalence of proteinuira and enzymuria among a cohort of subjects exposed to hexachlorobutadiene (HCBD) in their homes and to determine whether there was a change in observed effects when exposure ceased. Residents underwent a health check, which
Hexachloro-1,3-butadiene: toxicology, metabolism, and mechanisms of toxicity.
R S Yang
Reviews of environmental contamination and toxicology, 101, 121-137 (1988-01-01)
Arianna Chiusolo et al.
Toxicology letters, 182(1-3), 84-90 (2008-09-23)
Gene expression of regucalcin (Rgn), a calcium-binding protein, was investigated in kidney of male Wistar rats treated with proximal tubule segment-specific nephrotoxicants, namely hexachloro-1:3-butadiene (HCBD), specific for S(3) segment (pars recta) and potassium dichromate (chromate) specific for S(1)-S(2) segments (pars
Patrizia Cristofori et al.
Journal of applied toxicology : JAT, 33(3), 183-189 (2011-09-14)
The present research investigates the occurrence of hyaline droplet (HD) accumulation related to age, dose and time after treatment in male Wistar rats given a single i.p. injection of hexachloro-1:3-butadiene (HCBD). In the study on age, rats from 1 to
Andrea Trevisan et al.
Journal of applied toxicology : JAT, 25(1), 13-19 (2005-01-26)
Renal tissue biomarkers (glutamine synthetase and p-aminohippuric acid uptake) were studied in male and female rats after treatment with hexachloro-1,3-butadiene. Reduced glutathione content also was also determined in liver and kidney. Histopathological examination (light microscopy) was then performed. The aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service