Skip to Content
Merck
All Photos(1)

Documents

47799

Supelco

Dibromomethane

analytical standard

Synonym(s):

Methylene bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Br2
CAS Number:
Molecular Weight:
173.83
Beilstein:
969143
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

vapor density

6.05 (vs air)

vapor pressure

34.9 mmHg ( 20 °C)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.541 (lit.)

bp

96-98 °C (lit.)

mp

−52 °C (lit.)

density

2.477 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

BrCBr

InChI

1S/CH2Br2/c2-1-3/h1H2

InChI key

FJBFPHVGVWTDIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dibromomethane may be used as an analytical reference standard for the quantification of the analyte in environmental samples and organic compounds using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A simple and sensitive method for the determination of volatile halogenated organic compounds in sea water in the amol l? 1 to pmol l? 1 range
Ekdahl A and Abrahamsson K
Analytica Chimica Acta, 357(3), 197-209 (1997)
Determination of non-metals in organic compounds by gas chromatography with a miniature capacitively coupled plasma emission detector
Guchardi R and Hauser.CP
Journal of Analytical Atomic Spectrometry, 19(8), 945-949 (2004)
Marie E Archbold et al.
Rapid communications in mass spectrometry : RCM, 19(3), 337-342 (2005-01-13)
A pre-concentration system has been validated for use with a gas chromatography/mass spectrometry/isotope ratio mass spectrometer (GC/MS/IRMS) to determine ambient air (13)C/(12)C ratios for methyl halides (MeCl and MeBr) and chlorofluorocarbons (CFCs). The isotopic composition of specific compounds can provide
D A Whittington et al.
Biochemistry, 40(12), 3476-3482 (2001-04-12)
To investigate the role of protein cavities in facilitating movement of the substrates, methane and dioxygen, in the soluble methane monooxygenase hydroxylase (MMOH), we determined the X-ray structures of MMOH from Methylococcus capsulatus (Bath) cocrystallized with dibromomethane or iodoethane, or
Glenn A Marsch et al.
Chemical research in toxicology, 17(1), 45-54 (2004-01-20)
The dihalomethane CH(2)Cl(2) is an industrial solvent of potential concern to humans because of its potential genotoxicity and carcinogenicity. To characterize DNA damage by dihalomethanes, a rapid DNA digestion under acidic conditions was developed to identify alkali labile DNA-dihalomethane nucleoside

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service