Skip to Content
Merck
All Photos(1)

Key Documents

47527-U

Supelco

2-Isopropyl-3-methoxypyrazine solution

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
77101502
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-30°C

SMILES string

n1c(c(ncc1)C(C)C)OC

InChI

1S/C8H12N2O/c1-6(2)7-8(11-3)10-5-4-9-7/h4-6H,1-3H3

InChI key

NTOPKICPEQUPPH-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile substances from actinomycetes: their role in the odor pollution of water.
N N Gerber
CRC critical reviews in microbiology, 7(3), 191-214 (1979-11-01)
Profile: What is that smell?
Erik Vance
Nature, 455(7214), 726-728 (2008-10-10)
G J Pickering et al.
Journal of food science, 72(7), S468-S472 (2007-11-13)
2-Isopropyl-3-methoxypyrazine (IPMP) is a grape-derived component of wine flavor in some wine varieties as well as the causal compound of the off-flavor known as ladybug taint (LBT), which occurs when Harmonia axyridis beetles are incorporated with the grapes during juice
Susanne Kögel et al.
Journal of chemical ecology, 38(7), 854-864 (2012-06-01)
Food type can affect all functional aspects of an insect's life. We investigated the effects of different diet regimes on life history parameters of the ladybird beetle Harmonia axyridis. Furthermore, we tested the importance of elytral color, sex, and diet
G J Pickering et al.
Journal of food science, 73(6), S262-S266 (2009-02-26)
2-isopropyl-3-methoxypyrazine (IPMP) is the compound responsible for the off-flavor known as ladybug taint, which occurs when Harmnonia axyridis beetles become incorporated with the grapes during juice processing. It is also an important grape-derived component of juice flavor in some varieties.

Protocols

Separation of 2-Isopropyl-3-methoxypyrazine 20 ppt; 2-Isobutyl-3-methoxypyrazine; 2-Methylisoborneol; (±)-Geosmin 10 ppt

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service