28689-U
Carbotrap® T420 Thermal Desorption Tube
glass, O.D. x I.D. x L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm), fritted, preconditioned, pkg of 10 tubes
Synonym(s):
Carbotrap® T420
About This Item
Recommended Products
Product Name
Carbotrap® T420 89mm Glass-Fritted TD Tube, O.D. × L 1/4 in. (6.35 mm) × 3 1/2 in. (89 mm), preconditioned, pkg of 10 tubes
material
glass TD tube
Quality Level
Agency
meets requirements for US EPA/CLP TO-17
description
For use with PerkinElmer™, Markes, DANI, Shimadzu™, GL Science, and O-I Analytical thermal desorbers
product line
Carbotrap®
feature
fritted
preconditioned
packaging
pkg of 10 tubes
greener alternative product characteristics
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
technique(s)
active air sampling: suitable
O.D. × L
1/4 in. (6.35 mm) × 3 1/2 in. (89 mm)
matrix
Proprietary
compatibility
for analyte group Terpenes
greener alternative category
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Air Monitoring
- Environmental
- Industrial Hygiene
- Terpene Flavor Profiling
- Terpene Fragrance Profiling
- Terpene Odors
Storage and Stability
Legal Information
accessory
analytical column
recommended
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Discover our new Carbotrap® T420 tube, designed for precise terpene air sampling in the cannabis and hemp industry. Monitor terpenes in cultivation environments, even in high humidity, with reproducible sample recovery. Elevate your analysis with the latest in terpene sampling technology.
Discover our new Carbotrap® T420 tube, designed for precise terpene air sampling in the cannabis and hemp industry. Monitor terpenes in cultivation environments, even in high humidity, with reproducible sample recovery. Elevate your analysis with the latest in terpene sampling technology.
Discover our new Carbotrap® T420 tube, designed for precise terpene air sampling in the cannabis and hemp industry. Monitor terpenes in cultivation environments, even in high humidity, with reproducible sample recovery. Elevate your analysis with the latest in terpene sampling technology.
Discover our new Carbotrap® T420 tube, designed for precise terpene air sampling in the cannabis and hemp industry. Monitor terpenes in cultivation environments, even in high humidity, with reproducible sample recovery. Elevate your analysis with the latest in terpene sampling technology.
Protocols
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service