Skip to Content
Merck
All Photos(2)

Key Documents

21014

Supelco

LpDNPH S10 Cartridge

bed wt. 350 mg, volume 3 mL, pkg of 50 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000

Agency

ASTM® D5197
EPA 100,IP-6A,TO-11A
NIOSH 2016

Quality Level

composition

bed wt., 350 mg
2 4-DNPH (High-purity silica gel)

packaging

pkg of 50 ea

technique(s)

active air sampling: suitable

volume

3 mL

application(s)

air monitoring
environmental
industrial hygiene

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The design of the S10 cartridge makes it easy to use in the field and laboratory. Reusable adaptors are available for connecting the cartridge to the sampling pump. The built in reservoir eliminates the need to attach a syringe for sample extraction/elution.

Application

LpDNPH S10 Cartridge was suitable as sampling apparatus for aldehydes while performing gas chromatography. It was also used to sample DNPH derivatives to perform HPLC and GC-MS to measure and characterize DNOH derivatives of aldehydes and ketone.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kuo-Pin Yu et al.
Journal of the Air & Waste Management Association (1995), 56(5), 666-674 (2006-06-03)
Nowadays, the heating, ventilation, and air conditioning (HVAC) system has been an important facility for maintaining indoor air quality. However, the primary function of typical HVAC systems is to control the temperature and humidity of the supply air. Most indoor
Mehrdad Arshadi et al.
The Annals of occupational hygiene, 53(8), 797-805 (2009-08-12)
Different qualities of biofuel pellets were made from pine and spruce sawdust according to an industrial experimental design. The fatty/resin acid compositions were determined by gas chromatography-mass spectrometry for both newly produced pellets and those after 2 and 4 weeks

Related Content

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service