Skip to Content
Merck
All Photos(7)

Key Documents

ZRB2279

Sigma-Aldrich

Anti-FGF-20 Antibody, clone 3I3-L2 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Fibroblast growth factor 20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
IHC
WB
affinity binding assay
clone:
3I3-L2, recombinant monoclonal
species reactivity:
human
citations:
technique(s):
affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3I3-L2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... FGF20(26281)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 3I3-L2 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects FGF-20. It targets an epitope within 24 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal half of human FGF-20.

Application

Quality Control Testing

Evaluated by Western Blotting in Human cerebral cortex membrane preparations.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected FGF-20 in Human cerebral cortex membrane preparations.

Quality Control Testing

Evaluated by Western Blotting in Human cerebral cortex membrane preparations.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected FGF-20 in Human cerebral cortex membrane preparations.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected FGF-20 in Human cerebral cortex tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound FGF-20 peptide with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Fibroblast growth factor 20 (UniProt: Q9NP95; also known as FGF-20) is encoded by the FGF20 gene (Gene ID: 26281) in human. FGF-20 is a homodimeric protein that serves as a neurotrophic factor and regulates the development and function of central nervous system. It is predominantly expressed in the cerebellum and substantia nigra. Human and rat FGF-20 display high degree of homology (~ 95%). It acts on proximal cells and significantly enhances the survival of dopaminergic neurons by activating the MAPK pathway through FGF receptor 1c. In the rat model of Parkinson′s disease, FGF-20 is shown to protect against the loss of dopaminergic neurons. It is shown to ACT® synergistically with FGF-2 to increase the number of dopaminergic neurons in primate embryonic stem cell–derived neurospheres composed of neural progenitors. Exposure to FGF-20, along with Wnt5a and FGF-2, during the final stage of in vitro differentiation, can enhance the maturation and in vivo survival of dopaminergic neurons. Mutations in FGF20 gene have been linked to Renal hypodysplasia/aplasia 2 (RHDA2), a perinatally lethal renal disease with kidney development defects, bilateral renal aplasia, hypoplasia, and severe obstructive uropathy. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Itoh, N., and Ohta, H. (2013). Front. Mol. Neurosci. 6; 15; Ohmachi, S., et al. (2000). Biochem. Biophys. Res. Commun. 277(2); 355-360).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ACT is a registered trademark of Merck KGaA, Darmstadt, Germany
Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service