Skip to Content
Merck
All Photos(7)

Key Documents

ZRB1856

Sigma-Aldrich

Anti-14-3-3 β/α Antibody, clone 1D15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D15, monoclonal
recombinant monoclonal

description

1D15 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~28.08 kDa
observed mol wt ~28 kDa

species reactivity

human, monkey

species reactivity (predicted by homology)

sheep, rat, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1D15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects 14-3-3 protein β/a. It targets an epitope within 19 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal half of human 14-3-3 protein β/a.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected 14-3-3 β/a in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected 14-3-3 β/a in SHSY5Y and COS-1 cell lysates.

ELISA Analysis: 1.0 μg/mL from a representative lot detected recombinant 14-3-3 β/a.

Affinity Binding Assay: A representative lot of this antibody bound 14-3-3 b/a peptide with a KD of 2.3 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-14-3-3 β/a, clone 1D15 ZooMAb®, Cat. No. ZRB1856, is a recombinant Rabbit monoclonal antibody that detects 14-3-3 β/a and is tested for use in Affinity Binding Assay, ELISA, and Western Blotting.

Target description

14-3-3 protein β/a (UniProt: P31946; also known as Protein 1054, Protein kinase C inhibitor protein 1, KCIP-1) is encoded by the YWHAB gene (Gene ID: 7529) in human. Mammals express seven distinct 14-3-3 isoforms (gamma, epsilon, beta, zeta, sigma, theta, tau) that form multiple homo- and heterodimers, thus rendering 14-3-3 as a key multifunctional regulatory molecule. 14-3-3 protein beta/alpha serves as an adapter protein that has been implicated in the regulation of a large spectrum of both general and specialized signaling pathways. It binds to many partners, usually by recognition of a phosphoserine or phosphothreonine motif and its binding generally results in the modulation of the activity of the binding partner. The alpha, brain-specific form, differs from the beta form in its ability to be phosphorylated on serine 60 by protein kinase C delta in a sphingosine-dependent fashion. 14-3-3 β/a is shown to block the nuclear translocation of the phosphorylated form of SR protein-specific kinase 2 (SRPK2) and antagonize its stimulatory effect on cyclin D1 expression that results in blockage of SRPK2 elicited neuronal apoptosis. It also reported to serve as a negative regulator of osteogenesis. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service