Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1814

Sigma-Aldrich

Anti-ADAR1 Antibody, clone 1I4-H1 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
ICC
IHC (p)
WB
affinity binding assay
clone:
1I4-H1, recombinant monoclonal
species reactivity:
human, rat
citations:
technique(s):
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I4-H1, recombinant monoclonal

description

1I4-H1 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 136.01 kDa
observed mol wt ~150 and 110 kDa

purified by

using Protein A

species reactivity

human, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ADAR(103)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 114-H1 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Double-stranded RNA-specific adenosine deaminase (ADAR1). It targets an epitope within the internal region.

Immunogen

His-tagged recombinant fragment corresponding to 243 amino acids from the internal region of human Double-stranded RNA-specific adenosine deaminase (ADAR1).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ADAR1 in HeLa cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ADAR1 in NTERA-2 and L6 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ADAR1 in human cerebral cortex tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound ADAR1 with a KD of 3.4 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lo detected ADAR1 in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Double-stranded RNA-specific adenosine deaminase (UniProt: P55265; also known as EC:3.5.4.37, DRADA, 136 kDa double-stranded RNA-binding protein, p136, Interferon-inducible protein 4, IFI-4, K88DSRBP, ADAR1) is encoded by the ADAR (also known as ADAR1, DSRAD, G1P1, IFI4) gene (Gene ID: 103) in human. In mammals three different ADAR proteins have been described: ADAR1, -2, and -3. ADAR1 is a ubiquitously expressed homodimeric enzyme that catalyzes the hydrolytic deamination of adenosine to inosine (A to I) in double-stranded RNA (dsRNA). A-to-I editing is shown to predominantly occur in noncoding, repetitive elements and short interspersed nuclear elements (SINEs). This A-to-I editing affects gene expression and function in several ways, including mRNA translation by changing codons and hence the amino acid sequence of proteins, RNA stability by changing sequences during viral RNA replication, and RNA structure-dependent activities such as microRNA production or targeting or protein-RNA interactions. Homodimerization of ADAR1 is shown to be essential for its catalytic activity. Five isoforms of ADAR1 have been described that are produced by alternative promoter usage and alternative splicing. Isoform 5, which can form heterodimers with ADAR2, is expressed at higher levels in astrocytomas and its expression increases with severity of the tumor. Loss of ADAR1 is shown to overcome resistance to PD-1 checkpoint blockade caused by inactivation of antigen presentation by tumor cells and sensitizes tumors to interferons. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ishizuka, JJ., et al. (2019). Nature. 565(7737); 43-48; Cho, DSC., et al. (2003). J. Biol. Chem. 278(19); 17093-17102; George, CX., and Samuel, CE (1999). Proc. Natl. Acad. Sci. USA. 96(8); 4621-4626).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service