Skip to Content
Merck

Skip To

ZRB1663

Anti-Histone H2A.X Antibody, clone 1C22 ZooMAb® Rabbit Monoclonal

greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

H2a/x, Histone H2AX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
recombinant monoclonal
Application:
FACS, ICC, IHC (p), WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 15.15 kDa, observed mol wt ~16 kDa

species reactivity

mouse, human, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Histone H2AX (UniProt: P16104; also known as H2a/x, Histone H2A.X) is encoded by the H2AX (also known as H2AFX) gene (gene ID: 3014) in human. Histones are highly conserved basic nuclear proteins that are responsible for the nucleosome structure of chromatin in eukaryotes. They play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which DNA is wrapped in repeating units, called nucleosomes, which limits DNA accessibility to the cellular machineries that require DNA as a template. The histone H2A.X is a variant member of the H2A family of histones that is distinguished from other H2A histones by a unique carboxy-terminal sequence. This unique sequence is highly conserved throughout eukaryotic evolution and is rapidly phosphorylated by ATM or ATR at serine 139 in mammals in response to DNA double-strand breaks. H2A.X phosphorylation is important in the formation of a stable repair complex at the site of DNA damage. Phosphorylation of H2A.X is important in the formation of a stable repair complex at the site of DNA damage. This phosphorylation is a very rapid response to DNA damage, occurring within as little as one minute after exposure to ionizing radiation. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Immunogen

KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal region of human Histone H2AX.

Application

Quality Control Testing
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Histone H2A.X in MCF-7 cell lysate.Tested applicationsWestern Blotting Analysis: A 1:1,000 dilution from a representative lot detected Histone H2A.X in C6, Neuro-2A, and PC12 cell lysates.Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Histone H2A.X in HeLa, A431, HepG2, NIH 3T3, AC16, and MCF-7 cells.Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected Histone H2A.X in human cerebral cortex tissue sections.Flow Cytometry Analysis: 1 μg from a representative lot detected Histone H2A.X in one million HeLa cells.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Histone H2A.X, clone 1C22 ZooMAb, Cat. No. ZRB1663, is a recombinant Rabbit monoclonal antibody that targets Histone H2A.X and is tested for use in Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Biochem/physiol Actions

Clone 1C22 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Histone H2AX. It targets an epitope within 15 amino acids from the C-terminal region.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Preparation Note

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.
Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ZRB1624ZRB1134ZRB04791
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit (recombinant)

clone

recombinant monoclonal

clone

4O11, monoclonal, recombinant monoclonal

clone

recombinant monoclonal

clone

MC315, recombinant monoclonal

species reactivity

mouse, human, rat

species reactivity

human, rat, monkey, mouse

species reactivity

mouse, human, rat

species reactivity

human, mouse

UniProt accession no.

P16104

UniProt accession no.

Q8N257

UniProt accession no.

P37840

UniProt accession no.

Q16695


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters