Skip to Content
Merck
All Photos(29)

Key Documents

ZRB1147

Sigma-Aldrich

Anti-Calnexin Antibody, clone 10N19, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-CNX, Anti-IP90, Anti-P90

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

10N19, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 67.57 kDa
observed mol wt ~78 kDa

species reactivity

human, mouse

species reactivity (predicted by homology)

rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CANX(821)
mouse ... Canx(12330)
rat ... Canx(29144)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 10N19 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Calnexin. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human Calnexin.

Application

Anti-Calnexin, clone 10N19 ZooMAb , Cat. No. ZRB1147, is a highly specific rabbit recombinant monoclonal antibody that targets Calnexin and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin). and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Calnexin in Jurkat, NIH/3T3, and MCF7 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Calnexin in human tonsil and mouse testis tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Calnexin in HeLa, A431, HUVEC, NIH 3T3, Jurkat and MCF-7 cells.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Calnexin (UniProt: P27824; also known as IP90, Major histocompatibility complex class I antigen-binding protein p88, p90) is encoded by the CANX gene (Gene ID: 821) in human. Calnexin is a member of the calreticulin family of proteins. It is a well-conserved, single-pass type I membrane protein that serves as calcium binding, endoplasmic reticulum (ER) resident chaperone protein. It associates with newly synthesized, incompletely folded monomeric glycoproteins and promotes their correct folding prior to secretion. It is also reported to target misfolded proteins for degradation. It is anchored to the ER membrane by a transmembrane domain. Calnexin associates with partial T-cell antigen receptor complexes that escape the ER of immature thymocytes and may, therefore, function as a signaling complex regulating thymocyte maturation. Calnexin is synthesized with a signal peptide of 20 amino acids, which is subsequently cleaved off to generate the mature form that has a lumenal domain (aa 21-481), a short transmembrane domain (aa 482-502) and a cytoplasmic domain (aa 503-592). Three isoforms of calnexin have been described that are generated by alternative splicing. Calnexin is shown to be phosphorylated at serine 564 by MAPK3/ERK1, which increases its association with ribosomes. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service