ZMS1130
Anti-GABRB2 Antibody, clone BD17 ZooMAb® Mouse Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Gamma-aminobutyric acid receptor subunit beta-2;GABA(A) receptor subunit beta-2
About This Item
Recommended Products
biological source
mouse
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
BD17, recombinant monoclonal
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 54.45 kDa
observed mol wt ~52 kDa
purified by
using protein G
species reactivity
human, mouse
species reactivity (predicted by homology)
bovine, feline, rat, monkey
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
bovine ... GABRB2(540657)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Mouse liver membrane preparation.
Western Blotting Analysis (WB): A 1:1,000 dilution of this antibody detected GABRB2 in Mouse liver membrane preparation.
Tested Applications
Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected GABRB2 in Human brain and Mouse brain tissue sections.
Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected GABRB2 in Human cerebellum tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service