Key Documents
Z740421
Tamper proof specimen containers
size 500 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
polypropylene cap
polypropylene container
packaging
pack of 100 ea
manufacturer/tradename
Simport C581-500
size
500 mL
General description
These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples. Tight-fitting lids prevent leaks and odors. They incorporate a tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover. They resist temperatures up to 80 °C. The two larger sizes are supplied with a plastic handle for easy carrying.
Choose from one of the most recent versions:
Certificates of Analysis (COA)