Z732346
IKA® ULTRA-TURRAX® disperser tools, stainless steel
dispersing element for T-25 digital disperser, 8 mm, 1/cs
Synonym(s):
disperser blades, tissue homogenizers
All Photos(2)
About This Item
UNSPSC Code:
41103800
NACRES:
NB.22
Recommended Products
material
stainless steel
feature
stainless steel rotor/stator
packaging
pack of 1
manufacturer/tradename
IKA 0001024200
diam.
8 mm
Related Categories
General description
IKA® S 25 N - 8 G Dispersing tool for use with T 25 digital and easy cleabn ULTRA-TURRAX® dispersing instruments.
Features and Benefits
- suitable for use with solvents
- sterilization methods: all methods
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)