Z710962
Rack for glass slides
for 20/40 slides, glass
Synonym(s):
900304
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
Recommended Products
material
glass
feature
Holds 20/40 x slides 76x26 mm
manufacturer/tradename
WHEATON® 900304
size
118x88x70 mm
General description
Glass rack holds 20 single slides, 39 slides arranged alternately straight across and diagonally, or 40 slides back to back. Manufactured from soda-lime glass.
Features and Benefits
- Manufactured from soda-lime glass
- These mix-and-match components offer greater flexibility in meeting your requirements
- This staining dish accommodates 16, 20 and 30 slide racks
- It holds slides sizes 3” x 1” (75mm x 25mm), 3” x 1-1/2” (75mm x 38mm), and 3” x 2” (75mm x 51mm)
- Approximate inside bottom dimensions (mm): 121 L x 90 W x 66 D
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)