Key Documents
Z709093
ART® non-filtered tip
HLT 1000 SoftFit, size 100-1000 μL, Pure packed, sterile
Synonym(s):
pipet tips, pipette tips
About This Item
UNSPSC Code:
41121600
eCl@ss:
32079201
NACRES:
NB.01
maximum volume:
1 mL
size:
100-1000 μL
sterility:
sterile
feature:
DNase free
RNase free
Pure packed
packaging:
pack of 800 ea
manufacturer/tradename:
Molecular BioProducts 3581
sterility
sterile
feature
DNase free
RNase free
Pure packed
packaging
pack of 800 ea
manufacturer/tradename
Molecular BioProducts 3581
maximum volume
1 mL
size
100-1000 μL
General description
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
SoftFit-L design makes loading and ejecting easy and provides a universal fit.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)