Z673420
KDS stainless steel syringes
volume 50 mL, pressure 700-1500 psi
Synonym(s):
stainless steel syringes
All Photos(1)
About This Item
Recommended Products
description
with Luer lock
packaging
pack of 1 ea
manufacturer/tradename
KD Scientific 780817
parameter
700-1500 psi pressure
L × O.D.
5.49 in. (139.3 mm) × 1.25 in. (31.8 mm)
volume
50 mL
General description
These rugged #316 stainless steel syringes are a perfect complement to the KDS 410 high pressure syringe dispenser. They eliminate the problem of breaking glass syringes when using high pressures. Compatibility with most chemicals is assured as wetted parts are #316 stainless steel, Viton® for the 20, 50 and 100 mL sizes, and Perfluoroelastomer for the 8 mL size.
Features and Benefits
- Compatible with most syringe pumps
- Fully autoclavable
- Good resistance to most chemicals
- Safe solution when using high pressure dispensing
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company