Z640395
Eppendorf® epT.I.P.S. Reloads
volume range 0.1-20 μL
Synonym(s):
epT.I.P.S. Reload System, pipetting tips
About This Item
Eppendorf® 022491512
Recommended Products
material
medium gray tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 (10 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030 073.380
Eppendorf® 022491512
L
40 mm
maximum volume
20 μL
volume range
0.1-20 μL
volume
0.1—20 μL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately)
Application
Features and Benefits
- Reduced waste in comparison to disposable racks
- Refill trays can be autoclaved for reuse, without foil
- Packaging for the refill system varies based on tip size, including dual-sided or stacked options
- Color-coded trays enable easy identification of volume and pipettes
- Single-use pipette tips with a universal cone for compatibility
- User-friendly and easy to handle
- Offered in various purity grades
- Volume marking allows for convenient visual inspection
- Simple attachment and ejection with minimal force required
- Complete sealing ensures reliable performance
- Provides maximum precision and accuracy
- Long pipette tips are available for specific volume sizes
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service