Documents
Z332259
Bottle adapters for BRAND® Dispensers and Burettes
GL 32 thread, bottle (NS 19/26), polypropylene
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
material
polypropylene
manufacturer/tradename
BRAND 704419
fitting
bottle (NS 19/26)
GL 32 thread
Related Categories
General description
For all Dispensette® models as well as seripettor® and QuikSip
Other Notes
BR704419 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG