XNAB2RE
Extract-N-Amp™ Blood PCR Kit
sufficient for 1000 extractions, sufficient for 5000 amplifications
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
usage
sufficient for 1000 extractions
sufficient for 5000 amplifications
color
colorless
shipped in
wet ice
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
The Extract-N-Amp™ Blood PCR Kits contain all of the reagents necessary to rapidly extract host genomic DNA from whole blood and amplify targets of interest by direct PCR. This novel extraction system eliminates the need for any type of purification, organic extraction, centrifugation, heating, filtration or alcohol precipitation. The kit also includes a PCR ReadyMix™, specially formulated for amplification directly from the extract. This formulation uses an antibody based hot start, for specific amplification.
Application
Extract-N-Amp™ Blood PCR Kit has been used to extract DNA from genomes and dried blood spot samples. It has also been used in polymerase chain reaction (PCR).
Features and Benefits
- Efficient 8 minute prep allows greater speed and throughput
- No need for any type of purification, organic extraction, centrifugation or alcohol precipitation
- Simple, 3 step procedure with no special equipment required
- Hot start antibody included for highly specific PCR amplification of genomic DNA
- Compatible with multiple format (single tube or 96-well plate)
- Can be used with whole blood or blood cards
- Extract stable at 4 °C for at least 6 months
Principle
Genomic DNA is extracted from 10 μl of whole blood by simply adding the Extraction Solution and incubating for 5 minutes at room temperature. The Neutralization Solution is added to the extract to counteract inhibitory substances prior to PCR. A portion of the DNA extract is then added to the specially formulated PCR ReadyMix.
Other Notes
For additional information, please see www.sigma-aldrich.com/extract-n-amp.
Use of this product is covered by one or more of the following US patents and corresponding patent claims outside the US: 5,789,224, 5,618,711, 6,127,155 and claims outside the US corresponding to expired US Patent No. 5,079,352. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patents require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Legal Information
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser′s own internal research. No other patent rights (such as 5′ Nuclease Process patent rights) are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.
Antibody licensed for in vitro research use under U.S. Patent No. 5,338,671 and 5,587,287, and corresponding patents in other countries.
Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Electrophoresis, 30(14), 2532-2535 (2009-07-30)
Stored surplus of dried blood spot (DBS) samples from neonatal screening programs constitute a vast potential for large genetic epidemiological studies. However, age of the samples and the small amounts of DNA available may limit their usage. In this study
BioTechniques, 45(4), 423-424 (2008-10-16)
Sodium bisulfite treatment followed by PCR and DNA sequencing is widely considered the gold standard for the analysis of DNA methylation patterns. However, this technique generally requires substantial quantities of genomic DNA as starting material and is often associated with
Cancer research, 67(13), 6417-6424 (2007-07-10)
Idiopathic myelofibrosis (IM) is likely the consequence of both the acquisition of genetic mutations and epigenetic changes that silence critical genes that control cell proliferation, differentiation, and apoptosis. We have explored the effects of the sequential treatment with the DNA
BMC genomics, 10, 297-297 (2009-07-07)
Identification of disease susceptible genes requires access to DNA from numerous well-characterised subjects. Archived residual dried blood spot samples from national newborn screening programs may provide DNA from entire populations and medical registries the corresponding clinical information. The amount of
Endocrinology, 144(7), 2865-2869 (2003-06-18)
Transgenic mice overexpressing leptin backcrossed to the C57BL/6J genetic background (LepTg) have a lean phenotype, characterized by a 95% reduction in adipose mass; reduced plasma levels of glucose, triglycerides, insulin, and IGF-1; and a 75% decrease in adipocyte size. High-fat
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service