The Instructions for Use and the SDS offer some basic information regarding the formulation for Wright Stain WS32. However, the actual production formulation is considered proprietary.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
solution
Quality Segment
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
concentration
0.3%
technique(s)
microbe id | staining: suitable
pH
6.75-6.95 (25 °C)
application(s)
hematology
histology
storage temp.
room temp
General description
Application
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| technique(s) microbe id | staining: suitable | technique(s) - | technique(s) microbe id | staining: suitable | technique(s) - |
| concentration 0.3% | concentration 0.3% | concentration 0.4 % (w/v) in methanol (buffered at pH 6.8) | concentration 0.4 % (w/v) in methanol (buffered at pH 6.8) |
| form solution | form solution | form solution | form solution |
| application(s) hematology | application(s) hematology | application(s) hematology | application(s) hematology |
| Quality Level 500 | Quality Level 500 | Quality Level 500 | Quality Level 500 |
| shelf life Expiry date on the label. | shelf life Expiry date on the label. | shelf life Expiry date on the label. | shelf life Expiry date on the label. |
Still not finding the right product?
Explore all of our products under Wright Stain, Modified
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 1
target_organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
91.4 °F - closed cup
flash_point_c
33 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is it possible to provide the actual chemical ingredient list of the Wright stains?
1 answer-
Helpful?
-





