Skip to Content
Merck
All Photos(3)

Key Documents

WH0149233M1

Sigma-Aldrich

Monoclonal Anti-IL23R antibody produced in mouse

clone 3D7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-interleukin 23 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3D7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL23R(149233)

General description

Interleukin 23 receptor (IL23R) gene codes for a subunit of the IL-23 receptor. The gene is mapped to human chromosome 1p31.3.

Immunogen

IL23R (NP_653302, 553 a.a. ~ 628 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LNQGECSSPDIQNSVEEETTMLLENDSPSETIPEQTLLPDEFVSCLGIVNEELPSINTYFPQNILESHFNRISLLE

Biochem/physiol Actions

Interleukin 23 receptor (IL23R) IL-12Rβ1 and forms IL-23 complex, which is essential for IL-23 signaling. It constitutively combines with Janus kinase 2 (JAK2). IL23R attaches to transcription activator signal transducer and activator of transcription 3 (STAT3) in a ligand-dependent manner and has a proinflammatory function. IL23R gene defends against Crohn′s disease.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuhisa Mizuki et al.
Nature genetics, 42(8), 703-706 (2010-07-14)
Behçet's disease is a chronic systemic inflammatory disorder characterized by four major manifestations: recurrent ocular symptoms, oral and genital ulcers and skin lesions. We conducted a genome-wide association study in a Japanese cohort including 612 individuals with Behçet's disease and
Anthony W Segal
European journal of clinical investigation, 48 Suppl 2, e12983-e12983 (2018-06-23)
Crohn's disease (CD) is caused by a trigger, almost certainly enteric infection by one of a multitude of organisms that allows faeces access to the tissues, at which stage the response of individuals predisposed to CD is abnormal. In CD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service