Skip to Content
Merck
All Photos(4)

Documents

WH0010618M2

Sigma-Aldrich

Monoclonal Anti-TGOLN2 antibody produced in mouse

clone 2F11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-MGC14722, Anti-TGN38, Anti-TGN46, Anti-TGN48, Anti-TGN51, Anti-TTGN2, Anti-trans-golgi network protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2F11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TGOLN2(10618)

General description

Trans-Golgi network integral membrane protein 2 (TGOLN2) is a heterodimeric type I integral membrane protein. It has a highly conserved N terminus which consists of a signal peptide. The C terminus comprises part of the lumenal domain, a membrane spanning region and cytoplasmic tail.

Immunogen

TGOLN2 (NP_006455, 229 a.a. ~ 327 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
QGPIDGPSKSGAEEQTSKDSPNKVVPEQPSRKDHSKPISNPSDNKELPKADTNQLADKGKLSPHAFKTESGEETDLISPPQEEVKSSEPTEDVEPKEAE

Biochem/physiol Actions

Trans-Golgi network integral membrane protein 2 (TGOLN2) cycles between the trans-Golgi network (TGN) and the cell surface via an early endosomal compartment. This movement is mediated by a tyrosine-based tetra peptide signal (SDYQRL) in the cytoplasmic domain. It plays an important role in the formation of exocytic vesicles at the TGN by functioning as a receptor for complexes of a cytoplasmic protein known as p62 and one GTP-binding protein. The cytoplasmic domain of TGOLN2 binds to the complex and is essential for budding to occur. Coupling of the segregation of secretory proteins to the budding of exocytic vesicles is mediated by it. The cytosolic domain of TGOLN2 interacts with AP2 clathrin adaptor complexes and also with the coiled coil region of a protein called neurabin.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct interaction of the trans-Golgi network membrane protein, TGN38, with the F-actin binding protein, neurabin.
Stephens DJ and Banting G
The Journal of Biological Chemistry, 274(42), 30080-30086 (1999)
TGN38/41: a molecule on the move.
Stanley KK and Howell KE
Trends in Cell Biology, 3(8), 252-255 (1993)
Primate homologues of rat TGN38: primary structure, expression and functional implications.
Ponnambalam S
Journal of Cell Science, 675-685 (1996)
K K Stanley et al.
Trends in cell biology, 3(8), 252-255 (1993-08-01)
TGN38/41 is a heterodimeric integral membrane protein that cycles between the trans Golgi network and the cell surface. A tyrosine-containing tetrapeptide motif within its cytoplasmic tail is necessary and sufficient for determining its steady-state location in the TGN. Recent results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service