Skip to Content
Merck
All Photos(3)

Key Documents

WH0006855M1

Sigma-Aldrich

Monoclonal Anti-SYP antibody produced in mouse

clone 3B3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-synaptophysin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B3, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SYP(6855)

Immunogen

SYP (AAH64550.1, 1 a.a. ~ 313 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLLLADMDVVNQLVAGGQFRVVKEPLGFVKVLQWVFAIFAFATCGSYSGELQLNVDCANKTESDLSIEVEFEYPFRLHQVYFDAPTCRGGTTKVFLVGDYSSSAEFFVTVAVFAFLYSMGALATYIFLQNKYRENNKGPMLDFLATAVFAFMWLVSSSAWAKGLSDVKMATDPENIIKEMPVCRQTGNTCKELRDLVTSGLNTSVVFGFLNLVLWVGNLWFVFKETGWAAPFLRAPPGAPEKQPAPGDAYGDAGYGQGPGGYGPQDSYGPQGGYQPDYGQPAGSGGSGYGPQGDYGQQGYGPQGAPTSFSNQM

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuji Suzuki et al.
Digestive surgery, 31(2), 143-150 (2014-06-28)
The aim of this study was to determine the clinicopathological features and surgical management of solid pseudopapillary neoplasms (SPNs) of the pancreas at a single institution. We investigated 34 patients (5 males and 29 females) who underwent surgery for pathologically
Taiji Tsunemi et al.
Human molecular genetics, 23(11), 2791-2801 (2013-12-18)
Mutations in ATP13A2 (PARK9) cause Kufor-Rakeb syndrome (KRS) characterized by juvenile-onset parkinsonism, pyramidal signs and dementia. PARK9 belongs to type 5 P-type ATPase with its putative function as a cation transporter. Loss of PARK9 leads to lysosomal dysfunction and subsequent
Carlos B Mantilla et al.
Journal of applied physiology (Bethesda, Md. : 1985), 117(8), 910-920 (2014-08-30)
Activation of the tropomyosin-related kinase receptor B (TrkB) by brain-derived neurotrophic factor acutely regulates synaptic transmission at adult neuromuscular junctions (NMJs). The role of TrkB kinase activity in the maintenance of NMJ function and structure at diaphragm muscle NMJs was
Huiling Qu et al.
Restorative neurology and neuroscience, 32(5), 597-609 (2014-07-19)
The mechanism underlying forced limb-use -induced structural plasticity remains to be studied. We examined whether the cyclic adenosine monophosphate (cAMP)-mediated signal transduction pathway was involved in brain plasticity and promoted behavioral recovery induced by forced limb-use after stroke. Adult rats
Antonella Ferrante et al.
Neurobiology of disease, 71, 193-204 (2014-08-19)
Adenosine A1 receptor (A1R) stimulation exerts beneficial effects in response to various insults to the brain and, although it was found neuroprotective in a lesional model of Huntington's disease (HD), the features of this receptor in genetic models of HD

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service