Skip to Content
Merck
All Photos(6)

Key Documents

WH0000174M1

Sigma-Aldrich

Monoclonal Anti-AFP antibody produced in mouse

clone 1G7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FETA, Anti-HPAFP, Anti-alpha-fetoprotein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1G7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgGκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AFP(174)

General description

This gene encodes alpha-fetoprotein, a major plasma protein produced by the yolk sac and the liver during fetal life. Alpha-fetoprotein expression in adults is often associated with hepatoma or teratoma. However, hereditary persistance of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the alpha-fetoprotein and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. Alpha-fetoprotein is found in monomeric as well as dimeric and trimeric forms, and binds copper, nickel, fatty acids and bilirubin. The level of alpha-fetoprotein in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. (provided by RefSeq)

Immunogen

AFP (AAH27881, 500 a.a. ~ 609 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
CCTSSYANRRPCFSSLVVDETYVPPAFSDDKFIFHKDLCQAQGVALQTMKQEFLINLVKQKPQITEEQLEAVIADFSGLLEKCCQGQEQEVCFAEEGQKLISKTRAALGV

Application

Monoclonal Anti-AFP antibody produced in mouse has been used in immunocytochemistry.

Biochem/physiol Actions

Alpha-fetoprotein encoded by the gene AFP is involved in the regulation of fatty acids in both fetal and proliferating adult liver cells. Its expression is found to be increased in acute liver injuries, indicating active liver regeneration. It has been associated with fatty liver disease (FLD), a disease that may lead to cirrhosis and hepatocellular carcinoma. It serves as a tumor marker for HCC (hepatocellular carcinoma).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carla Sanjurjo-Soriano et al.
Stem cell research, 33, 247-250 (2018-11-24)
We generated an induced pluripotent stem cell (iPSC) line using dermal fibroblasts from a patient with Usher syndrome type 2 (USH2). This individual was homozygous for the most prevalent variant reported in the USH2A gene, c.2299delG localized in exon 13.
Anika Neureiter et al.
Stem cell research, 33, 20-24 (2018-10-09)
Angelman syndrome (AS) is a neurodevelopmental disorder with leading symptoms of happy demeanor, intellectual disability, ataxia and seizures. AS can be caused by genetic and epigenetic aberrations, resulting in the absence of functional UBE3A protein in the brain. UBE3A is
Björn Lichtner et al.
Stem cell research, 15(3), 697-699 (2016-03-19)
Primary human chorionic villi (CV) cells were used to generate the iPSC line by retroviral transduction of the four Yamanaka-factors OCT4, SOX2, KLF4 and c-MYC. Pluripotency was confirmed both in vivo and in vitro. The transcriptomes of the CV-derived iPSC
Julia Tigges et al.
ALTEX, 38(4), 595-614 (2021-05-09)
Human induced pluripotent stem cells (hiPSC) are a promising tool for replacing animal-based experiments. To warrant data reproducibility, quality-controlled research material is recommended. While the need for global harmonization of quality standards for stem cell banking centers, commercial providers, pre-clinical
Gergő Vőfély et al.
Molecular and cellular neurosciences, 88, 222-230 (2018-02-10)
Pluripotent stem cell derived human neuronal progenitor cells (hPSC-NPCs) and their mature neuronal cell culture derivatives may efficiently be used for central nervous system (CNS) drug screening, including the investigation of ligand-induced calcium signalization. We have established hippocampal NPC cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service