Skip to Content
Merck
All Photos(1)

Documents

V2264

Sigma-Aldrich

Vinorelbine ditartrate salt hydrate

≥98% (HPLC), powder

Synonym(s):

3′,4′-Didehydro-4′-deoxy-C′-norvincaleukoblastine [R-(R*,R*)-2-3-dihydroxybutanedioate (1:2)salt], 5′-Noranhydrovinoblastine tartrate, KW-2307, NVB, Navelbine tartrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H54N4O8 · 2C4H6O6 · xH2O
CAS Number:
Molecular Weight:
1079.11 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

off-white

solubility

H2O: 10 mg/mL

originator

Pierre Fabre

shipped in

wet ice

storage temp.

2-8°C

SMILES string

[H]O[H].O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.[H][C@]12C[N@H](CC(CC)=C1)Cc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C)[C@@]7([H])[C@](O)([C@H](OC(C)=O)[C@]8(CC)C=CCN9CC[C@]67[C@]89[H])C(=O)OC

InChI

1S/C45H54N4O8.2C4H6O6.H2O/c1-8-27-19-28-22-44(40(51)55-6,36-30(25-48(23-27)24-28)29-13-10-11-14-33(29)46-36)32-20-31-34(21-35(32)54-5)47(4)38-43(31)16-18-49-17-12-15-42(9-2,37(43)49)39(57-26(3)50)45(38,53)41(52)56-7;2*5-1(3(7)8)2(6)4(9)10;/h10-15,19-21,28,37-39,46,53H,8-9,16-18,22-25H2,1-7H3;2*1-2,5-6H,(H,7,8)(H,9,10);1H2/t28-,37-,38+,39+,42+,43+,44-,45-;2*1-,2-;/m011./s1

InChI key

ZLIPKSVXTCRHIB-GNBHHJMASA-N

Biochem/physiol Actions

Vinorelbine is a potent anti-mitotic, anti-tumor agent. Vinorelbine inhibits microtubule assembly. Low neurotoxicity is related to its higher affinity for mitotic microtubules than for axonal microtubules.

Features and Benefits

This compound was developed by Pierre Fabre. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lihui Wang et al.
Cell death & disease, 8(4), e2746-e2746 (2017-04-14)
Chemotherapeutic agents are generally used as a frontline therapy for non-small cell lung cancer (NSCLC). However, resistance to chemotherapy arises rapidly in NSCLC, and the reasons for chemotherapy resistance have not been fully determined. Here, we found cisplatin, but not
Hongyang Li et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 89, 1277-1285 (2017-03-23)
Vinorelbine (NVB) is a semi-synthetic vinca alkaloid that is approved for the clinical therapy of lung cancer. However, the clinical application of NVB was limited because of the acquisition of resistance and inacceptable toxicity. Therefore, it is of great interest
Fabio Castiglione et al.
Current urology reports, 18(11), 90-90 (2017-10-02)
Due to the increasing numbers of radical prostatectomies (RP) performed for prostate cancer, a substantial number of patients are now suffering from post-operative erectile dysfunction (ED). The aim of this study is to summarize the current literature on surgical techniques
Juliana Teixeira Fiquer et al.
Journal of affective disorders, 238, 636-644 (2018-06-30)
It is unclear if individuals with Major Depressive Disorder (MDD) present different nonverbal behavior (NVB) compared with healthy individuals, and also if depression treatments affect NVB. In this study, we compared the NVB of MDD subjects and healthy controls. We
Man Su Kim et al.
Journal of microbiology (Seoul, Korea), 57(5), 405-412 (2019-02-24)
Xanthorrhizol (XTZ), isolated from Curcuma xanthorrhiza, has potent antifungal and antibacterial activity. It shows very strong activity against Gram-positive bacteria, such as Streptococcus mutans and Staphylococcus aureus, but is generally not active against Gram-negative bacteria. In this study, we explored

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service