Skip to Content
Merck
All Photos(1)

Key Documents

V1253

Sigma-Aldrich

Anti-Vascular Endothelial Growth Factor antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-VEGF

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

capture ELISA: suitable
immunohistochemistry: suitable
neutralization: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Vegfa(22339)

General description

Vascular Endothelial Growth Factor (VEGF) is a 46kD dimeric, heparin binding glycoprotein. It facilitates several functions like induction of endothelial cell proliferation, inhibits apoptosis, induces permeabilization of blood vessels and promotes cell migration. Anti-Vascular Endothelial Growth Factor antibody can be used for neutralizing the biological activity of recombinant mouse (VEGF164) and recombinant mouse (VEGF120). It can also be used in immunoblotting. Goat anti-Vascular Endothelial Growth Factor antibody reacts specifically with recombinant mouse VEGF.

Specificity

The antibody will neutralize the biological activity of recombinant mouse VEGF164, and recombinant mouse VEGF121. It can also be used as a capture antibody in mouse VEGF 164 or 120 sandwich ELISAs.

Immunogen

purified 164 amino acid residue variant of recombinant mouse VEGF.

Application

Anti-Vascular Endothelial Growth Factor antibody can be used in sandwich ELISAs as a capture antibody in mouse VEGF 164 or 120. It can also be used in immunohistochemistry.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Merve Nur Eğlenen et al.
Turkish journal of orthodontics, 36(2), 79-86 (2023-06-22)
Functional treatment of Class II malocclusion is expected to lead to adaptation in the condyle. This study aimed to evaluate the effects of adipose tissue-derived mesenchymal stem cells (ADMSCs), low-level laser therapy (LLLT), and grape-seed extract (GSE) on condylar growth
Dorien Reijnders et al.
Physiological genomics, 50(5), 385-392 (2018-03-10)
Preeclampsia (PE), a hypertensive disease of pregnancy, is a leading cause of fetal and maternal morbidity/mortality. Early angiogenic and inflammatory disturbances within the placenta are thought to underlie the development of the maternal PE syndrome and poor pregnancy outcomes. However
Johan Dixelius et al.
Cancer research, 66(4), 2089-2097 (2006-02-21)
Histidine-rich glycoprotein (HRGP) is an abundant heparin-binding plasma protein that efficiently arrests growth and vascularization of mouse tumor models. We have shown that the antiangiogenic effect of HRGP is dependent on its histidine/proline-rich domain, which needs to be released from
Vascular endothelial growth factor induces cyclooxygenase-dependent proliferation of endothelial cells via the VEGF-2 receptor.
J F Murphy et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 15(9), 1667-1669 (2001-06-28)
Hui Wang et al.
Cell cycle (Georgetown, Tex.), 17(17), 2164-2174 (2018-09-11)
The highly malignant feature and difficulties for early diagnosis are the key reasons contributing to the poor prognosis of pancreatic cancer (PC) patients. NFE2L3 is a nuclear transcription factor, which has been reported an important biomarker of several tumors. But

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service