Skip to Content
Merck
All Photos(1)

Key Documents

V113

Sigma-Aldrich

S(+)-γ-Vigabatrin

solid

Synonym(s):

S(+)-γ-Vinyl GABA, S(+)-4-Aminohexenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

optical activity

[α]/D +11.8°, c = 2.5 in H2O(lit.)

color

off-white

solubility

H2O: >20 mg/mL
ethanol: soluble

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(O)=O)C=C

InChI

1S/C6H11NO2/c1-2-5(7)3-4-6(8)9/h2,5H,1,3-4,7H2,(H,8,9)/t5-/m1/s1

InChI key

PJDFLNIOAUIZSL-RXMQYKEDSA-N

Application

S(+)-γ-Vigabatrin has been used as a standard in ultra-high-performance-liquid-chromatography (UHPLC).

Biochem/physiol Actions

γ-Vigabatrin has the potential to treat infantile spasms and seizures. It exhibits therapeutic effects against cocaine and other addictions.
GABA transaminase inhibitor; anti-convulsant.

Caution

Hygroscopic

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluating risks for vigabatrin treatment
Krauss, Gregory L
Epilepsy currents, 9(5), 125-129 (2009)
Determination of enantiomeric vigabatrin by derivatization with diacetyl-l-tartaric anhydride followed by ultra-high performance liquid chromatography-quadrupole-time-of-flight mass spectrometry
Zhao J, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 1040(5), 199-207 (2017)
O M Larsson et al.
Neuropharmacology, 25(6), 617-625 (1986-06-01)
A preferential effect of valproate on gamma-aminobutyric acid (GABA) in the nerve terminal compartment has been proposed. Gamma-vinyl GABA, an irreversible inhibitor of GABA-transaminase (GABA-T) causes a preferential increase in the GABA compartment of the non-nerve terminal. The aim of
M M Robin et al.
European journal of pharmacology, 62(4), 319-327 (1980-04-04)
Unilateral intrastriatal injection of various substances induces a characteristic dyskinetic syndrome in rats. These substances include picrotoxin as well as a series of irreversible GABA-transaminase inhibitors. Using the degree of enzyme inhibition in various brain areas as a measure of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service