Skip to Content
Merck
All Photos(1)

Key Documents

UC126

Sigma-Aldrich

4-Hydroxymephenytoin

Synonym(s):

(±)-5-Ethyl-5-(4-hydroxyphenyl)-3-methylhydantoin, (±)-5-Ethyl-5-(4-hydroxyphenyl)-3-methylimididazolidine-2,4-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O3
CAS Number:
Molecular Weight:
234.25
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

mp

153-154 °C

storage temp.

2-8°C

SMILES string

CCC1(NC(=O)N(C)C1=O)c2ccc(O)cc2

InChI

1S/C12H14N2O3/c1-3-12(8-4-6-9(15)7-5-8)10(16)14(2)11(17)13-12/h4-7,15H,3H2,1-2H3,(H,13,17)

InChI key

OQPLORUDZLXXPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CYP2C19 metabolite of mephenytoin.

Application

4-Hydroxymephenytoin has been used for developing ultra performance LC (UPLC)- tandem mass spectrometry (MS/MS) assays for evaluating cytochrome P450 (CYP) probe drugs and their relevant metabolites in human urine or plasma samples. 4-hydroxymephenytoin has also been used for developing sub-2-micron particle LC/MS/MS-based methods for analyzing CYP450 probe substrate metabolites.

Biochem/physiol Actions

4-Hydroxymephenytoin catabolism displays genetic polymorphism and is a direct indicator of Cytochrome P450 2C19 (CYP2C19) activity. Detection and quantification of 4-hydroxymephenytoin in biological samples by HPLC method is well optimized.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simple and selective assay of 4-hydroxymephenytoin in human urine using solid-phase extraction and high-performance liquid chromatography with electrochemical detection and its preliminary application to phenotyping test
Tanaka M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 676(1), 87-94 (1996)
High-performance liquid chromatographic determination of urinary 4?-hydroxymephenytoin, a metabolic marker for the hepatic enzyme CYP2C19, in humans
Xie HG, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 668(1), 125-131 (1995)
Nicolas A Stewart et al.
The Analyst, 136(3), 605-612 (2010-11-26)
The efficiency of drug metabolism by a single enzyme can be measured as the fractional metabolic clearance which can be used as a measure of whole body activity for that enzyme. Measurement of activity of multiple enzymes simultaneously is feasible
C Desiderio et al.
Electrophoresis, 15(1), 87-93 (1994-01-01)
Using cyclodextrin micellar electrokinetic capillary chromatography (CD-MECC), baseline separation of mephenytoin, 4-hydroxymephenytoin and 4-hydroxyphenytoin enantiomers in urine was effected with beta-cyclodextrin. After single-dose administration of 100 mg of racemic mephenytoin, the 0-8 h urine was collected, and enzymatically hydrolyzed urine
A Adedoyin et al.
Clinical pharmacology and therapeutics, 64(1), 8-17 (1998-08-08)
Drug metabolism is influenced by liver disease because of the central role that the liver plays in metabolic activities in the body. However, it is still unclear how activities of specific drug-metabolizing enzymes are influenced by the presence and severity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service