Skip to Content
Merck

U4635

UNISETTE Tissue Processing/Embedding Cassettes

white

Synonym(s):

UNISETTE Tissue Processing/Embedding Cassettes

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.17
UNSPSC Code:
41116124
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


color

white

application(s)

hematology
histology

storage temp.

room temp

General description

Histology processing/embedding cassettes made of acetal polymer. These cassettes keep specimens safely submerged in liquid and are resistant to histological solvents. The one-piece lid snaps apart and conveniently locks into the cassette base, ensuring specimen safety. A line of cassette colors are available to fit your needs and preference. A large 30° anterior writing area is available for your convenience. Not suitable for automated printers.

Application

Compatible with Leica and Sakura printers.

Preparation Note

Case contains 3 dispenser boxes of 500 cassettes.

Legal Information

Unisette is a trademark of Simport


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
U4635-1CS04061826763544