Skip to Content
Merck

U3510

Universal Antibody Dilution Buffer, Ready-To-Use, with BSA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
12352107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


pH

7.3-7.7

Quality Segment

application(s)

hematology
histology

storage temp.

room temp

General description

Ready-to-use buffer for dilution of antibodies in all immunoassays. This buffer contains immunoglobulin free BSA and does not contain phosphate, sodium azide or mercury preservatives. It can be used for dilution of all antibodies, including peroxidase conjugates and antibodies to phosphoproteins. Not suitable for dilution of antibodies to S100 proteins.

Application

For dilution of primary or secondary antibodies. The diluted antibodies can be stored at 2-8 °C for several weeks.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library