Skip to Content
Merck
All Photos(1)

Key Documents

U2875

Sigma-Aldrich

Uric acid sodium salt

Synonym(s):

2,6,8-Trihydroxypurine, Sodium acid urate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H3N4O3Na
CAS Number:
Molecular Weight:
190.09
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

microbial
synthetic

Assay

11.0-13.0% anhydrous basis (Sodium, ICP)

form

powder

solubility

1 M NaOH: 25 mg/mL, clear, colorless to faintly yellow

SMILES string

[Na].O=C1NC(=O)C2=C(N1)NC(=O)N2

InChI

1S/C5H4N4O3.Na.H/c10-3-1-2(7-4(11)6-1)8-5(12)9-3;;/h(H4,6,7,8,9,10,11,12);;

InChI key

IHMJZGFANDMEJM-UHFFFAOYSA-N

Application

Uric acid sodium salt has been used in isotonic solution of NaCl dose titration study. It has been used as a suspension in NaCl for intra-articular administration to experimental animals to induce synovitis.

Biochem/physiol Actions

Sodium urate is the sodium salt of uric acid. In gout, it is observed in synovial fluid. Sodium urate has been shown to precipitate calcium oxalate from urine. It is also known to reverse the inhibitory effects of reactive oxygen and nitrogen metabolites on gelatinase A activity. Excess sodium urate in soft tissues results in a condition referred to as tophi.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dissolved urate salts out calcium oxalate in undiluted human urine in vitro: implications for calcium oxalate stone genesis
Grover PK, et al.
Chemistry & Biology, 10(3), 271-278 (2003)
Kim Gilbert et al.
The journal of trauma and acute care surgery, 86(3), 431-439 (2018-12-07)
Multiple organ failure can develop after hemorrhagic shock (HS). Uric acid (UA) is released from dying cells and can be proinflammatory. We hypothesized that UA could be an alternative mediator of organ apoptosis and inflammation after HS. Ventilated male Wistar
Medical Biochemistry, 410-410 (2014)
Rocío Cánovas et al.
Lab on a chip, 17(14), 2500-2507 (2017-06-28)
A novel low-cost, compact and sensitive paper-based platform for the accurate monitoring of glucose in biological fluids is presented. Paper-based working and reference electrodes are combined to build a whole potentiometric cell, which also fits a sampling module for simple
Preferential accumulation of meloxicam in inflamed synovial joints of dogs
Johnston L and Narbe R
The Veterinary Record, vetrec-ve2011 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service