Skip to Content
Merck

TMO144530

Nunc® Non-Treated Multidish

wells 24, polystyrene, L × W 128 mm × 86 mm, with lid, non-treated, case of 75 ea

Synonym(s):

multiwell plate, non treated multiwell plate, non treated plate, Nunc 144530, cultivation plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.13
UNSPSC Code:
41161502
Material:
polystyrene
Manufacturer/tradename:
Nunc 144530
Packaging:
case of 75 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

packaging

case of 75 ea

manufacturer/tradename

Nunc 144530

L × W

128 mm × 86 mm

wells

24

General description

Perform suspension cell cultures with Thermo Scientific Nunc Non-Treated Multidishes.

Application

  • Suspension cell cultures.

Features and Benefits

  • Flat-bottom round wells.
  • Excellent optical properties.
  • Raised well rims to lower the risk of cross-contamination.
  • Lid with notched corners for good fit and easy orientation.
  • Sterile and non-pyrogenic.

Legal Information

Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?

Explore all of our products under Nunc® Non-Treated Multidish




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library