All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C23H48O
CAS Number:
Molecular Weight:
340.63
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
storage temp.
2-8°C
Quality Level
SMILES string
CCCCCCCCCCCCCCCCCCCCCCCO
InChI
1S/C23H48O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24/h24H,2-23H2,1H3
InChI key
FPLNRAYTBIFSFW-UHFFFAOYSA-N
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Saet Buyl Lee et al.
Plant & cell physiology, 57(11), 2300-2311 (2016-09-01)
Aerial plant surfaces are coated by a cuticular wax layer to protect against environmental stresses, such as desiccation. In this study, we investigated the functional relationship between MYB94 and MYB96 transcription factors involved in cuticular wax biosynthesis. Both MYB94 and
Josep Massana-Codina et al.
Frontiers in plant science, 11, 1287-1287 (2020-09-26)
Black dot is a blemish disease of potato tubers caused by the phytopathogenic fungus Colletotrichum coccodes. Qualitative resistance (monogenic) that leads to the hypersensitive response has not been reported against black dot, but commercial potato cultivars show different susceptibility levels
Jennifer Bragg et al.
TAG. Theoretical and applied genetics. Theoretische und angewandte Genetik, 133(11), 3119-3137 (2020-08-18)
Quantitation of leaf surface wax on a population of switchgrass identified three significant QTL present across six environments that contribute to leaf glaucousness and wax composition and that show complex genetic × environmental (G × E) interactions. The C4 perennial
Sol Ji Choi et al.
Food chemistry, 204, 94-101 (2016-03-19)
Policosanol (PC) is a mixture of health promoting bioactive long-chain aliphatic alcohols. Here, we report that green tea (Camellia sinensis) leaves are the exceptionally rich plant-sources of PC. Young and tender leaves and old and turf leaves of C. sinensis
Hyojin Kim et al.
Plant cell reports, 36(6), 815-827 (2017-03-11)
An increased permeability of the cuticle is closely associated with downregulation of genes involved in cuticular lipid synthesis in hypoxia-stressed Arabidopsis and may allow plants to cope with oxygen deficiency. The hydrophobic cuticle layer consisting of cutin polyester and cuticular
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service