Skip to Content
Merck

T7204

Toremifene citrate salt

≥98% (HPLC)

Synonym(s):

(Z)-2-(4-(4-Chloro-1,2-diphenyl-1-butenyl)phenoxy)-N,N-dimethylethanamine, Acapodene, Fareston, GTx 006, Z-Toremifene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H28ClNO · C6H8O7
CAS Number:
Molecular Weight:
598.08
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
51111800
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CN(C)CCOc1ccc(cc1)C(=C(\CCCl)c2ccccc2)\c3ccccc3

InChI

1S/C26H28ClNO.C6H8O7/c1-28(2)19-20-29-24-15-13-23(14-16-24)26(22-11-7-4-8-12-22)25(17-18-27)21-9-5-3-6-10-21;7-3(8)1-6(13,5(11)12)2-4(9)10/h3-16H,17-20H2,1-2H3;13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/b26-25-;

InChI key

IWEQQRMGNVVKQW-OQKDUQJOSA-N

Gene Information

General description

Toremifene is a triphenylethylene derivative. It exhibits antitumor and antiestrogen effects. Toremifene has estrogen-agonist effects on blood lipids and endometrium.

Application

Toremifene citrate salt has been used:
  • in cell-based ELISA
  • as a positive control to detect its antiviral activity against Ebola virus (EBOV)
  • to treat MCF7 and T47D breast cancer cell lines

Biochem/physiol Actions

Toremifene citrate is an oral selective estrogen receptor modulator (SERM). It is used in advanced (metastatic) breast cancer and being evaluated for prevention of prostate cancer. Toremifene citrate is known to increase bone mineral density in prostate cancer patients undergoing androgen deprivation therapy†.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Preparation Note

Toremifene citrate salt is soluble in DMSO at a concentration that is greater than 10 mg/ml.


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library