Skip to Content
Merck
All Photos(1)

Key Documents

T6819

Sigma-Aldrich

Anti-phospho-Tau (pSer199/202) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DDPAC, Anti-FTDP-17, Anti-MAPTL, Anti-MSTD, Anti-MTBT1, Anti-MTBT2, Anti-PPND, Anti-PPP1R103, Anti-TAU

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

microarray: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer199/pSer202)

Gene Information

human ... MAPT(4137)
mouse ... Mapt(17762)
rat ... Mapt(29477)

General description

Tau (τ), also known as MAPT (microtubule associated protein tau), is encoded by the gene mapped to human chromosome 17q21.3. It is highly expressed in neurons, but is most prominent in axons.
Tau proteins are known as microtubule-associated phosphoprotein (MAP) and express mainly in neurons of central nervous system. It has a crucial role in tubulin polymerization and facilitates microtubules assembly and stability. The biological activity of tau proteins depends on degree of its phosphorylation. Anti-phospho-Tau (pSer199/202) antibody can be used in microarray and western blotting. It can also be used in immunoblotting. Rabbit anti- phospho-Tau (pSer199/202) antibody reacts specifically with human Tau (pSer199/202) (45-68 kD).

Immunogen

chemically synthesized phosphopeptide derived from the region of human tau that contains serine199 and serine202.

Application

Anti-phospho-Tau (pSer199/202) antibody produced in rabbit has been used in:
  • immunocytochemistry
  • immunohistochemistry
  • western blot
Anti-phospho-Tau (pSer199/202) antibody (diluted 1:1000 in PBS containing 0.3% Triton X-100 and 0.5% BSA) can be used in immunocytochemistry for identification of protein aggregates. It can also be used as primary antibody (diluted 1: 3000) in immunohistochemistry.

Biochem/physiol Actions

Tau (τ) plays an essential role in the assembly and maintenance of microtubule structure. Deletion of tau (τ) results in developmental delay and learning disability. The gene expression is associated with the development of Alzheimer′s disease (AD). Genetic variation in τ gene increases the risk of susceptibility to the sporadic tauopathies, progressive supranuclear palsy (PSP) and corticobasal degeneration.

Physical form

Solution in 100 μl Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, 50% glycerol, containing 1.0 mg/ml BSA (IgG, protease free) and 0.05% sodium azide. The amount of antibody is sufficient for 10 immunoblots.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

320.0 °F - closed cup

Flash Point(C)

160 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan-Bin Shi et al.
Frontiers in aging neuroscience, 12, 596894-596894 (2020-12-29)
Dystrophic neurites (DNs) are found in many neurological conditions such as traumatic brain injury and age-related neurodegenerative diseases. In Alzheimer's disease (AD) specifically, senile plaques containing silver-stained DNs were already described in the original literature defining this disease. These DNs
Feng-Qin Zhou et al.
Alzheimer's research & therapy, 10(1), 40-40 (2018-04-25)
Alzheimer's disease (AD) is a devastating neurodegenerative disorder bearing multiple pathological hallmarks suggestive of complex cellular/molecular interplay during pathogenesis. Transgenic mice and nonhuman primates are used as disease models for mechanistic and translational research into AD; the extent to which
Tian Tu et al.
Frontiers in aging neuroscience, 12, 93-93 (2020-06-02)
Amyloid plaques and neurofibrillary tangles (NFTs) are hallmark lesions of Alzheimer's disease (AD) related to β-amyloid (Aβ) deposition and intraneuronal phosphorylated tau (pTau) accumulation. Sortilin C-terminal fragments (shortened as "sorfra") can deposit as senile plaque-like lesions within AD brains. The
Mayuri Khandelwal et al.
Heliyon, 6(7), e04499-e04499 (2020-08-11)
Alzheimer's disease (AD) is the largest unmet medical complication. The devastation caused by the disease can be assumed from the disease symptoms like speech impairment, loss of self-awareness, acute memory loss etc. The individuals suffering from AD completely depend on
Rodrigo S Chaves et al.
BMC neuroscience, 11, 144-144 (2010-11-12)
Protein aggregates containing alpha-synuclein, beta-amyloid and hyperphosphorylated tau are commonly found during neurodegenerative processes which is often accompanied by the impairment of mitochondrial complex I respiratory chain and dysfunction of cellular systems of protein degradation. In view of this, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service