Skip to Content

T6789

Tris Buffered Saline, with BSA, pH 8.0

powder, pH 8.0, with BSA

Synonym(s):

1X TBS with 1% BSA, BSA blocking buffer, Tris Buffered Saline with 1% Bovine Serum Albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41105319
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tris Buffered Saline, with BSA, pH 8.0, powder

form

powder

Quality Segment

packaging

10 pak of (in foil bag)

color

off-white to yellow

pH

7.80—8.20

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

Preparation Note

Contents of one pouch, when dissolved in one liter of distilled or deionized water, will yield 0.05 M Tris buffered saline (NaCl - 0.138 M; KCl - 0.0027 M); bovine serum albumin - 1% (w/v), pH 8.0 at 25 °C.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library