Skip to Content

T5149

Microcentrifuge tubes with attached lid

volume 0.6 mL

Synonym(s):

Microcentrifuge Tubes, centrifuge tube, Simport Plastics T330-6N, sample collection tube, sample receptacle, sample tube

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.11
UNSPSC Code:
41121703
Material:
cap (Attached lid), clear , polypropylene
Sterility:
non-sterile
Packaging:
case of 1,000 ("Case of 1000), pack of 500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

cap (Attached lid), clear , polypropylene

sterility

non-sterile

packaging

case of 1,000 ("Case of 1000), pack of 500

manufacturer/tradename

Simport T330-6N

H × O.D.

30 mm × 8 mm

volume

0.6 mL

General description

Graduated rugged tubes made of laboratory-grade polypropylene suitable to withstand the stress of high-speed centrifugation up to 20,000g. The one-piece construction incorporates a snug-fitting and reliable attached cap even with prolonged boiling. The bottom is reinforced for added protection against leakage. Maximum clarity for visual sample inspection. A frosted writing surface on the closure and side of the tube allows for easy and convenient sample identification. A highly polished interior ensures low liquid retention. Packaged in tamperproof resealable safety-lock bags.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library