Skip to Content
Merck

Skip To

T4019

Trypsin−Agarose

buffered aqueous suspension, from bovine pancreas trypsin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352204
eCl@ss:
42020142
NACRES:
NA.54
MDL number:
Biological source:
bovine pancreas (trypsin)
Concentration:
≥15 units/mL (packed gel)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine pancreas (trypsin)

Quality Segment

form

buffered aqueous suspension

concentration

≥15 units/mL (packed gel)

extent of labeling

≥15 units per mL (packed gel)

matrix

beaded agarose

storage temp.

2-8°C

General description

The trypsin molecule has two domains: one is related to the enzyme active site and the tryptophan residues; the other is related to the 8-anilinonaphthalene-1-sulfonate binding.

Application

An experimental approach using SDS-PAGE, in-gel trypsin digestion and proteomic analysis identified five endosomal proteins. [1] Trypsin cleavage of the high-risk HPV-16 E5 protein (16E5) generates a unique four-amino acid C-terminal peptide that serves as a marker for E5 expression in transfected cells and epithelial cell lines containing integrated and episomal HPV-16 DNA. [2]

Physical form

Suspension in approx. 10 mM acetic acid, pH 3.2, containing preservative

Preparation Note

TPCK-treated

Other Notes

One unit will hydrolyze 1.0 μmole of BAEE per min at pH 8.0 at 30 °C (titrimetric assay).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T8802T6763T1763
biological source

bovine pancreas (trypsin)

biological source

bovine pancreas

biological source

bovine pancreas

biological source

bovine pancreas (trypsin)

concentration

≥15 units/mL (packed gel)

concentration

-

concentration

-

concentration

≥15 units/mL (packed gel)

form

buffered aqueous suspension

form

essentially salt-free, lyophilized powder

form

powder

form

buffered aqueous suspension

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

extent of labeling

≥15 units per mL (packed gel)

extent of labeling

-

extent of labeling

-

extent of labeling

≥15 units per mL packed gel


Still not finding the right product?

Explore all of our products under Trypsin−Agarose


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters