Skip to Content
Merck
All Photos(2)

Key Documents

T3766

Sigma-Aldrich

Thymine 1-β-D-arabinofuranoside

≥99%

Synonym(s):

araT, 1-β-D-Arabinofuranosylthymine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O6
CAS Number:
Molecular Weight:
258.23
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥99%

form

powder

optical activity

[α]20/D 90°, c = 5 mg/mL in water

solubility

0.5 M HCl: 20 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

CC1=CN(C2OC(CO)C(O)C2O)C(=O)NC1=O

InChI

1S/C10H14N2O6/c1-4-2-12(10(17)11-8(4)16)9-7(15)6(14)5(3-13)18-9/h2,5-7,9,13-15H,3H2,1H3,(H,11,16,17)

InChI key

DWRXFEITVBNRMK-UHFFFAOYSA-N

Application

1-β-D-Arabinofuranosylthymine (araT):
  • maybe used as a substrate and inhibitor to study the specificity and kinetics of thymidine kinase(s), such as herpes simplex virus type 1 and 2 thymidine kinases, the HSV1-tk and, HSV2-tk gene products.
  • is used to study the specificity and kinetics of various mitochondrial nucleoside kinases.
  • has been used as an antiviral agent to study its efficacy on Cryptosporidium parvum in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Balzarinia et al.
Biochemical pharmacology, 61(6), 727-732 (2001-03-27)
Introduction of a bulky lipophilic acyl entity at the 2'-OH position of both 1-beta-D-arabinofuranosylthymine (araT) and (E)-5-(2-bromovinyl)-1-beta-D-arabinofuranosyluracil (BVaraU), consistently resulted in a marked ( approximately 10-fold) increase in the inhibitory activity of these new arabinosyl nucleoside analogues for the mitochondrial
N Solaroli et al.
Antiviral research, 79(2), 128-132 (2008-05-06)
The thymidine kinases from feline herpesvirus (FHV TK) and canine herpesvirus (CHV TK) were cloned and characterized. The two proteins are closely sequence-related to each other and also to the herpes simplex virus type 1 thymidine kinase (HSV-1 TK). Although
Xiuyun Wang et al.
Nature communications, 11(1), 653-653 (2020-02-02)
The industrial synthesis of ammonia (NH3) using iron-based Haber-Bosch catalyst requires harsh reaction conditions. Developing advanced catalysts that perform well at mild conditions (<400 °C, <2 MPa) for industrial application is a long-term goal. Here we report a Co-N-C catalyst with high
Efficacy of select antivirals against Cryptosporidium parvum in vitro
Woods KM and Upton SJ
FEMS Microbiology Letters, 168(1), 59-63 (1998)
Kristof Y Neven et al.
Scientific reports, 10(1), 161-161 (2020-01-15)
Iodine is an essential trace element, necessary for the production of thyroid hormones, which play a key role in optimal foetal growth and (neuro-) development. To date, iodine deficiency remains a health burden in many countries. We investigated the variability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service