Skip to Content
Merck
All Photos(1)

Documents

SRP6507

Sigma-Aldrich

Haptoglobin (Phenotype 1-1) from human plasma

≥95% (SDS-PAGE)

Synonym(s):

Hp 1-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human plasma

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

86 kDa

packaging

pkg of 1 mg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HP(3240)

General description

Haptoglobin is a positive acute-phase plasma α2-sialoglycoprotein, and exists as three major phenotypes namely Hp 1-1, Hp 2-1, and Hp 2-2. Haptoglobin is composed of heavy β- (40 kDa) and light α- (α1= 8.9kDa and α2= 16kDa) chains. Hp 1-1 is the smallest phenotype with a molecular weight of 86kDa, and is composed of only α1.

Biochem/physiol Actions

Haptoglobin functions as an antioxidant by binding free hemoglobin, and eliminates it from circulation to prevent iron loss and renal injury following hemolysis. It is responsible for maintaining the helper T-cell type 1 and type 2 (Th1/Th2) balance, and functions as an immunosuppressor of lymphocyte function. It also functions as an angiogen and anti-inflammatory agent. Hp 1-1 phenotype is most effective in binding free hemoglobin and preventing inflammatory responses linked with free hemoglobin. Studies show that all the three phenotypes of haptoglobin are involved in idiopathic generalized epilepsy.

Physical form

Lyophilized as a salt-free solid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of haptoglobin genotype in an Iranian population with idiopathic generalized epilepsy.
Al-Balaghee S et al
Reports of Biochemistry and Molecular Biology, 3(2), 51-55 (2015)
Haptoglobin phenotypes in health and disorders.
Sadrzadeh SM, Bozorgmehr J, et al.
American Journal of Clinical Pathology, 121, S97-104 (2004)
Haptoglobin phenotypes in epilepsy.
Sadrzadeh SM, et al.
Clinical Chemistry, 50(6), 1095-1097 (2004)
Fabiana Baganha et al.
International journal of molecular sciences, 23(21) (2022-11-12)
Phosphorylcholine (PC) is one of the main oxLDL epitopes playing a central role in atherosclerosis, due to its atherogenic and proinflammatory effects. PC can be cleared by natural IgM antibodies and low levels of these antibodies have been associated with
Amrita Sharma et al.
iScience, 25(11), 105302-105302 (2022-10-29)
NEU-4438 is a lead for the development of drugs against Trypanosoma brucei, which causes human African trypanosomiasis. Optimized with phenotypic screening, targets of NEU-4438 are unknown. Herein, we present a cell perturbome workflow that compares NEU-4438's molecular modes of action

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service