Skip to Content
Merck
All Photos(1)

Key Documents

SRP6061

Sigma-Aldrich

VEGFR2 human

recombinant, expressed in insect cells, ≥95% (SDS-PAGE)

Synonym(s):

Fetal liver kinase 1, Kinase insert domain receptor, Protein-tyrosine kinase receptor flk-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in insect cells

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

116

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... VEGFR2(3791)

General description

VEGFR-2 has a lower affinity for VEGF than the Flt-1 receptor, but a higher signaling activity. Mitogenic activity in endothelial cells is mainly mediated by VEGFR-2 leading to their proliferation. No naturally occurring, secreted forms of VEGFR-2 have so far been reported. The binding of VEGF165 to VEGFR-2 is dependent on heparin. Soluble VEGFR-2 Human Recombinant produced in baculovirus is monomeric, glycosylated, polypeptide having a molecular mass of 116 kDa. The soluble receptor protein contains only the first 7 extracellular domains, which contain all the information necessary for ligand binding. The sKDR is purified by proprietary chromatographic techniques.
Vascular endothelial growth factor receptor 2 (VEGFR2), also known as kinase insert domain-containing receptor (KDR), is encoded by the gene mapped to human chromosome 4q12. The encoded protein belongs to the VEGF receptor family.

Biochem/physiol Actions

Vascular endothelial growth factor receptor 2 (VEGFR2) and its ligand VEGF play a vital role in angiogenesis and inflammation. It also facilitates autocrine stimulation of VEGF. VEGFR2 is implicated in progression of various types of cancers such as gliomas, malignant peripheral nerve sheath tumors, triple-negative breast cancer (TNBC) and gastrointestinal stromal tumors (GISTs).

Physical form

KDR was lyophilized from a sterile solution containing 25 mM MES pH 5.5 and 100 mM NaCl.

Preparation Note

Centrifuge the vial prior to opening.

Reconstitution

Reconstitute in sterile water to a concentration not less than 0.1 mg/mL. This solution can then be stored at 4 °C for 2-7 days. For future use for future use; For long term storage it is recommended to add a carrier protein (0.1 % HSA or BSA) then store at ?20 °C. Avoid freeze-thaw cycles.

Other Notes

ASVGLPSVSL DLPRLSIQKD ILTIKANTTL QITCRGQRDL DWLWPNNQSG SEQRVEVTEC SDGLFCKTLT IPKVIGNDTG AYKCFYRETD LASVIYVYVQ DYRSPFIASV SDQHGVVYIT ENKNKTVVIP CLGSISNLNV SLCARYPEKR FVPDGNRISW DSKKGFTIPS YMISYAGMVF CEAKINDESY QSIMYIVVVV GYRIYDVVLS PSHGIELSVG EKLVLNCTAR TELNVGIDFN WEYPSSKHQH KKLVNRDLKT QSGSEMKKFL STLTIDGVTR SDQGLYTCAA SSGLMTKKNS FVRVHEKPFV AFGSGMESLV EATVGERVRI PAKYLGYPPP EIKWYKNGIP LESNHTIKAG HVLTIMEVSE RDTGNYTVIL TNPISKEKQS HVVSLVVYVP TPQIGEKSLI SPVDSYQYGT TQTLTCTVYA IPPPHHIHWY WQLEEECANE PSQAVSVTNP YPCEEWRSVE DFQGGNKIEV NKNQFALIEG KNKTVSTLVI QAANVSALYK CEAVNKVGRG ERVISFHVTR GPEITLQPDM QPTEQESVSL WCTADRSTFE NLTWYKLGPQ PLPIHVGELP TPVCKNLDTL WKLNATMFSN STNDILIMEL KNASLQDQGD YVCLAQDRKT KKRHCVVRQL TVLERVAPTI TGNLENQTTS IGESIEVSCT ASGNPPPQIM WFKDNETLVE DSGIVLKDGN RNLTIRRVRK EDEGLYTCQA CSVLGCAKVE AFFIIEGA

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yung-Yi Chen et al.
Angiogenesis, 17(1), 207-219 (2013-10-17)
The binding of vascular endothelial growth factor (VEGF) to VEGF receptor-2 (VEGFR-2) on the surface of vascular endothelial cells stimulates many steps in the angiogenic pathway. Inhibition of this interaction is proving of value in moderating the neovascularization accompanying age-related
Zeng Li et al.
Iranian journal of basic medical sciences, 19(4), 411-416 (2016-06-10)
We previously reported a series of quinazoline derivatives as vascular-targeting anticancer agents. In this study, we investigated the mechanism underlying the anti-angiogenic activity of the quinazoline derivative compound 11d. We examined the effects of quinazoline derivative 11d: on vascular endothelial
Magdy A H Zahran et al.
Archiv der Pharmazie, 347(9), 642-649 (2014-06-20)
A series of thalidomide and phthalimide ester analogs were efficiently synthesized from N-chloromethylthalidomide, N-chloromethylphthalimide, and N-(2-bromoethyl)phthalimide derivatives with various biologically important carboxylic acids. The synthesized compounds were purified and characterized by various chromatographic and spectroscopic techniques. The antitumor activity of
Kristi D Lynn et al.
BMC cancer, 10, 397-397 (2010-08-03)
Vascular endothelial growth factor (VEGF) is a primary stimulant of angiogenesis under physiological and pathological conditions. Anti-VEGF therapy is a clinically proven strategy for the treatment of a variety of cancers including colon, breast, lung, and renal cell carcinoma. Since
Eunha Kim et al.
PloS one, 8(11), e81275-e81275 (2013-11-23)
The hepatocyte growth factor receptor (MET) is a receptor tyrosine kinase (RTK) that has emerged as an important cancer target. Consequently, a number of different inhibitors varying in specificity are currently in clinical development. However, to date, it has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service