Skip to Content
Merck
All Photos(1)

Key Documents

SRP5190

Sigma-Aldrich

HSP70, His tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HSP70-1, HSP72, HSPA1, HSPA1A, HSPA1B

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~70 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HSPA1A(3303)

Related Categories

General description

Heat shock protein 70 (HSP70) is a ubiquitous molecular chaperone. It comprises an N-terminal domain, nucleotide-binding domain (NBD), substrate-binding domain (SBD), and a C-terminal domain.

Application

Heat shock protein 70 (HSP70), His tagged human has been used:
  • to study the interaction between HSP70 and receptor of advanced glycation endproducts (RAGE) using protein proximity ligand assay (PLA)
  • to facilitate the import of superoxide dismutase 2 (SOD2) into the mitochondria
  • to study its role in muscle catabolism

Biochem/physiol Actions

Heat shock protein 70 (HSP70) plays a role in the cellular protein folding and remodeling process. It is also involved in the translocation of polypeptides into the chloroplast, mitochondria, and endoplasmic reticulum. HSP70 facilitates dismantling of protein complexes and modulates protein activity. It also plays a role in guarding cells from proteotoxic stress, pathophysiological conditions, and organismal aging. HSP70 participates in the activation of several immune cells such as macrophages, natural killer (NK) cells, B lymphocytes, peripheral monocytes, and antigen-presenting cells (APCs).

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of the stress-inducible human heat shock protein 70 substrate-binding domain in complex with peptide substrate.
Zhang P, et.al
PLoS ONE, 9(7), e103518-e103518 (2014)
Putative model for heat shock protein 70 complexation with receptor of advanced glycation end products through fluorescence proximity assays and normal mode analyses
Marcelo Sartori Grunwald
Cell Stress & Chaperones (2017)
Analysis of serum heat shock protein 70 (HSPA1A) concentrations for diagnosis and disease activity monitoring in patients with rheumatoid arthritis.
Cell Stress & Chaperones, 20(3) (2015)
Rina Rosenzweig et al.
Nature reviews. Molecular cell biology, 20(11), 665-680 (2019-06-30)
The 70-kDa heat shock proteins (Hsp70s) are ubiquitous molecular chaperones that act in a large variety of cellular protein folding and remodelling processes. They function virtually at all stages of the life of proteins from synthesis to degradation and are
H R Pelham
The EMBO journal, 3(13), 3095-3100 (1984-12-20)
The major heat-shock protein, hsp70, is synthesized by cells of many organisms in response to stress. In the present study, Drosophila hsp70 was expressed from cloned genes in mouse L cells and monkey COS cells and detected by immunofluorescence using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service