Skip to Content
Merck
All Photos(2)

Key Documents

SPC50120

Sigma-Aldrich

SP Sephadex® C-50 sodium form

Synonym(s):

Sulfopropyl Sephadex®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
NACRES:
NA.56
Pricing and availability is not currently available.

bead size

40-125 μm (dry)

Quality Level

pore size

200,000 exclusion limit (av. mol. wt.)

operating pH

2-10

capacity

2.0-2.8 meq/g ion exchange capacity
2.0-2.8 meq/g

compatibility

mode of use strong cation exchange chromatography

Application

SP Sephadex® is used in protein chromatography, ion exchange chromatography and cation exchange media. SP Sephadex® has been used to study human tumors as well as to discuss the biochemical basis for the use of mebendazole in the treatment of echinococcosis.

Legal Information

Sephadex is a registered trademark of Cytiva

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R De Prisco et al.
Biochimica et biophysica acta, 788(3), 356-363 (1984-08-14)
A ribonuclease, active on single- and double-stranded RNAs, has been isolated from human seminal plasma 3-5 micrograms of enzyme were recovered per ml of seminal plasma, equivalent to 71% of total activity and a 2500-fold purification (measured with poly(A) X
Isolation and partial characterization of a porcine thyroglobulin-binding <I>Phaseolus vulgaris</I> L. lectin.
Bonorden, W.R., and Swanson, B.G.
Food Chemistry, 44(3), 227-233 (1992)
B G Wallace
The Journal of neuroscience : the official journal of the Society for Neuroscience, 10(11), 3576-3582 (1990-11-01)
Heparin and heparan sulfate have been shown to block nerve-induced acetylcholine-receptor (AChR) aggregation at developing neuromuscular junctions. We found that heparin, heparan sulfate, and a wide variety of other polyanions also inhibited agrin-induced AChR aggregation. The more highly charged the
L Cheng et al.
The Journal of biological chemistry, 266(36), 24404-24412 (1991-12-25)
Rat cellular retinol-binding protein II (CRBP II) is an abundant 134-residue intestinal protein that binds all-trans-retinol and all-trans-retinal. It belongs to a family of homologous, 15-kDa cytoplasmic proteins that bind hydrophobic ligands in a noncovalent fashion. These binding proteins include
J B Starren et al.
Cellular immunology, 104(1), 59-70 (1987-01-01)
We have examined the isotypic pattern of the response of mice to the Ir gene-controlled antigen, dinitrophenyl-ovomucoid (DNP-OM). H-2 kappa mice are high responders (HR); (H-2b,d mice are low responders (LR). The isotype patterns of HR and LR strains differ

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service