Skip to Content
Merck

SML3967

Levonorgestrel

≥98% (HPLC)

Synonym(s):

(17α)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one, 13-Ethyl-17-ethynyl-17β-hydroxy-4-gonen-3-one, 13-Ethyl-17-hydroxy-18,19-dinor-17α-pregn-4-en-20-yn-3-one, D(−)-Norgestrel, D-Norgestrel, LNG

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -28 to -36° (C= 1.0g/100 ml in CDCl3)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O[C@@]1([C@@]2([C@H]([C@H]3[C@@H]([C@H]4CCC(=O)C=C4CC3)CC2)CC1)CC)C#C

InChI key

WWYNJERNGUHSAO-XUDSTZEESA-N

Biochem/physiol Actions

Orally bioavailable synthetic progestational agent that reversibly binds to progesterone receptors with high-affinity in the hypothalamus, pituitary gland, and uterus.

Levonorgestrel (D-Norgestrel) is an orally bioavailable synthetic progestational agent that reversibly binds to progesterone receptors with high-affinity in the hypothalamus, pituitary gland, and uterus. Levonorgestrel inhibits the release of gonadotropin-releasing hormone (GnRH), luteinizing hormone (LH), and follicle-stimulating hormone (FSH). D(−)-Norgestrel prevents fertilization by altering cervical mucus consistency, and interfering with the endometrial lining, making it less receptive to implantation. Levonorgestrel (LNG) can be used alone or in combination with estrogen during hormone replacement therapy.


Still not finding the right product?

Explore all of our products under Levonorgestrel


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Carc. 2 - Lact. - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library