Skip to Content
Merck
All Photos(1)

Key Documents

SML3900

Sigma-Aldrich

955, CDK9 PROTAC trifluoroacetate

≥98% (HPLC)

Synonym(s):

(E)-N-(5-(((5-(tert-butyl)oxazol-2-yl)methyl)thio)thiazol-2-yl)-1-(2-(4-(2-(2,6-dimethoxy-4-(3-oxo-3-(6-oxo-3,6-dihydropyridin-1(2H)-yl)prop-1-en-1-yl)phenoxy)ethyl)piperazin-1-yl)-2-oxoethyl)piperidine-4-carboxamide trifluoroacetate, PL-SNS-032 conjugate 955 trifluoroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H53N7O8S2 · xC2HF3O2
Molecular Weight:
836.03 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

955 CDK9 PROTAC is a cell penetrant conjugate of the CDK9 inhibitor SNS-032 with new recruiter of the E3 ligase KEAP1 piperlongumine that potently degrades CDK9 in MOLT4 cells. Similarly, to THAL-SNS-032, 955 induces CDK10 degradation. 955 exhibits potent antiproliferative activity in several prostate cancer cell lines including LNCaP, 22RV1, PC3 and DU145 cells.
Cell penetrant, potent and selective CDK9 degrader

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piperlongumine conjugates induce targeted protein degradation
Cell Chemical Biology, 30(2) (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service