Skip to Content
Merck

SML3833

NB-598

≥98% (HPLC)

Synonym(s):

(E)-3-[(3,3′-Bithiophen)-5-ylmethoxy]-N-(6,6-dimethyl-2-hepten-4-ynyl)- N-ethylbenzylamine, 3-([3,3′-Bithiophen]-5-ylmethoxy)-N-[(2E)-6,6-dimethyl-2-hepten-4-yn-1-yl]-N-ethylbenzenemethanamine, NB 598

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H31NOS2
CAS Number:
Molecular Weight:
449.67
UNSPSC Code:
51111800
NACRES:
NA.21
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

[s]1c(cc(c1)c3c[s]cc3)COc2cc(ccc2)CN(CC)C\C=C\C#CC(C)(C)C

InChI

1S/C27H31NOS2/c1-5-28(14-8-6-7-13-27(2,3)4)18-22-10-9-11-25(16-22)29-19-26-17-24(21-31-26)23-12-15-30-20-23/h6,8-12,15-17,20-21H,5,14,18-19H2,1-4H3/b8-6+

InChI key

KIRGLCXNEVICOG-SOFGYWHQSA-N

Biochem/physiol Actions

NB-598 is a potent and specific inhibitor of mammalian squalene epoxidase (SQLE) that suppresses triglyceride biosynthesis through the farnesol pathway. It decreases cholesterol levels in cells. NB-598 exhibit potent cytotoxic effects in subset of f neuroendocrine tumors, such as small cell lung cancers through accumulation of toxic squalene with in sensitive cancer cells.
Potent and specific inhibitor of mammalian squalene epoxidase (SQLE) that suppresses triglyceride biosynthesis through the farnesol pathway


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library