Skip to Content
Merck
All Photos(2)

Key Documents

SML3780

Sigma-Aldrich

Nintedanib ethanesulphonate

≥98% (HPLC)

Synonym(s):

(3Z)-2,3-Dihydro-3-[[[4-[methyl[2-(4-methyl-1-piperazinyl)acetyl]amino]phenyl]amino]phenylmethylene]-2-oxo-1H-indole-6-carboxylic acid methyl ester, monoethanesulphonate salt, (Z)-3-[(4-{Methyl-[2-(4-methylpiperazin-1-yl)acetyl]amino}phenylamino)methylene]-2-oxo-2,3-dihydro-1H-indole-6-carboxylic acid methyl ester, monoethanesulphonate salt, BIBF 1120 esylat, BIBF-1120 esylate, BIBF1120 esylate, Methyl (Z)-3-(((4-(N-methyl-2-(4-methylpiperazin-1-yl)acetamido)phenyl)amino)(phenyl)methylene)-2-oxoindoline-6-carboxylate, monoethanesulphonate salt, Nintedanib monoethanesulphonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H33N5O4·C2H6O3S
CAS Number:
Molecular Weight:
649.76
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

[S](=O)(=O)(O)CC.N5(CCN(CC5)C)CC(=O)N(C)c1ccc(cc1)N\C(=C3\c4c(cc(cc4)C(=O)OC)NC\3=O)\c2ccccc2

InChI

1S/C31H33N5O4.C2H6O3S/c1-34-15-17-36(18-16-34)20-27(37)35(2)24-12-10-23(11-13-24)32-29(21-7-5-4-6-8-21)28-25-14-9-22(31(39)40-3)19-26(25)33-30(28)38;1-2-6(3,4)5/h4-14,19,32H,15-18,20H2,1-3H3,(H,33,38);2H2,1H3,(H,3,4,5)/b29-28-;

InChI key

MMMVNAGRWOJNMW-FJBFXRHMSA-N

Biochem/physiol Actions

Nintedanib (BIBF1120) is an orally active, potent ATP-competitive inhibitor against angiokinases VEGFR-1/2/3 (IC50 = 34/21/13 nM), FGFR-1/2/3/4 (IC50 = 69/37/108/610 nM), PDGFRα/β (IC50 = 59/65 nM), as well as Flt-3, Lck, Lyn, and Src (IC50 = 26, 16, 195, 156 nM, repectively), but not 33 other kinases. Nintedanib exhibits antiangiogenic and antifibrotic efficacy in cultures and in animal models of cancers and pulmonary fibrosis in vivo.
Orally active, potent ATP-competitive VEGFR, FGFR, PDGFR, Flt3, Lck, Lyn, Src inhibitor with in vivo antiangiogenic and antifibrotic efficacy.

Caution

Hygroscopic

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B - STOT RE 1

Target Organs

Liver,Gastro-intestinal system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel Frenedoso da Silva et al.
Cell and tissue research, 379(2), 407-420 (2019-09-02)
The antiangiogenic therapy for prostate cancer with Nintedanib, a potent inhibitor of important growth factor receptors, has been proven to delay tumor progression and arrest tumor growth; thus, the aim herein is to evaluate Nintedanib effects on tumor cells, besides angiogenesis
Roman C Brands et al.
Oncology letters, 18(3), 2220-2231 (2019-08-28)
Multidrug resistance (MDR) remains one of the major causes of suboptimal outcome following therapy in head and neck squamous cell carcinoma (HNSCC). ATP-binding cassette (ABC) transporters are overexpressed in HNSCC, which contributes to the limited effect of chemotherapeutic treatment. In
Elaine Reguera-Nuñez et al.
Angiogenesis, 22(4), 535-546 (2019-08-30)
In contrast to VEGF pathway-targeting antibodies, antiangiogenic tyrosine kinase inhibitors (TKIs) have failed to meet primary endpoints in almost all phase III clinical trials when combined with conventional chemotherapy. One exception is the combination of nintedanib and docetaxel as a
Frank Hilberg et al.
Cancer research, 68(12), 4774-4782 (2008-06-19)
Inhibition of tumor angiogenesis through blockade of the vascular endothelial growth factor (VEGF) signaling pathway is a novel treatment modality in oncology. Preclinical findings suggest that long-term clinical outcomes may improve with blockade of additional proangiogenic receptor tyrosine kinases: platelet-derived
Gerald J Roth et al.
Journal of medicinal chemistry, 52(14), 4466-4480 (2009-06-16)
Inhibition of tumor angiogenesis through blockade of the vascular endothelial growth factor (VEGF) signaling pathway is a new treatment modality in oncology. Preclinical findings suggest that blockade of additional pro-angiogenic kinases, such as fibroblast and platelet-derived growth factor receptors (FGFR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service