Skip to Content
Merck

SML3779

Methotrexate disodium salt

≥98% (HPLC)

Synonym(s):

Amethopterin sodium, N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic acid disodium salt, Sodium methotrexate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H20N8Na2O5
CAS Number:
Molecular Weight:
498.40
UNSPSC Code:
12352200
NACRES:
NA.21
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to brown

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[Na].[Na].N([C@@H](CCC(=O)O)C(=O)O)C(=O)c1ccc(cc1)N(Cc2nc3c(nc(nc3N)N)nc2)C

InChI key

HQRCOVOSZPGUCA-GXKRWWSZSA-N

Biochem/physiol Actions

Folic acid antagonist that is used as an antineoplastic and antirheumatic drug
Methotrexate disodium salt is a potent is a folic acid antagonist that is used as an antineoplastic and antirheumatic drug. Methotrexate disodium salt is a potent inhibitor of dihydrofolate reductase.

Disclaimer

Hygroscopic


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library