Skip to Content
Merck

SML3731

Testosterone undecanoate

≥98% (HPLC)

Synonym(s):

(17β)-17-[(1-Oxoundecyl)oxy]androst-4-en-3-one, Undecanoic acid ester with testosterone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
UNSPSC Code:
51111800
NACRES:
NA.24
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

drug control

USDEA Schedule III

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

Biochem/physiol Actions

Testosterone undecanoate is an orally available prodrug of testosterone that has a very long elimination half-life. It is an androgen and anabolic–androgenic steroid that acts as agonist of the androgen receptor (AR). Testosterone undecanoate is mainly used in the treatment of low testosterone levels in men. Also, it was approved for treatment of male hypogonadism.
orally available prodrug of testosterone that has a very long elimination half-life; androgen that acts as agonist of the androgen receptor (AR)


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library