Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C23H23ClFNO5
CAS Number:
Molecular Weight:
447.88
UNSPSC Code:
51111800
NACRES:
NA.25
MDL number:
Quality Level
assay
≥98% (HPLC)
form
powder
optical activity
[α]/D -26 to -34°, c = 0.5 in methanol
color
white to beige
solubility
DMSO: 2 mg/mL, clear
storage temp.
-10 to -25°C
SMILES string
Fc1c(cccc1Cc2cc3c([n](cc([c]3=O)C(=O)O)[C@H](CO)C(C)C)cc2OC)Cl
InChI
1S/C23H23ClFNO5/c1-12(2)19(11-27)26-10-16(23(29)30)22(28)15-8-14(20(31-3)9-18(15)26)7-13-5-4-6-17(24)21(13)25/h4-6,8-10,12,19,27H,7,11H2,1-3H3,(H,29,30)/t19-/m1/s1
InChI key
JUZYLCPPVHEVSV-LJQANCHMSA-N
Biochem/physiol Actions
Elvitegravir (JTK-303; GS-9137) is an orally active human immunodeficiency virus type 1 (HIV-1) integrase inhibitor that prevents HIV-1 replication (EC50 = 0.9 nM using HTLV-I transformed T4-cell line MT-4) by blocking viral DNA to host DNA strand transfer (IC50 = 7.2 nM by cell-free assays).
Orally active human immunodeficiency virus type 1 (HIV-1) integrase strand transfer inhibitor.
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kazuya Shimura et al.
Journal of virology, 82(2), 764-774 (2007-11-06)
Integrase (IN), an essential enzyme of human immunodeficiency virus (HIV), is an attractive antiretroviral drug target. The antiviral activity and resistance profile in vitro of a novel IN inhibitor, elvitegravir (EVG) (also known as JTK-303/GS-9137), currently being developed for the
Anneleen Hombrouck et al.
Antimicrobial agents and chemotherapy, 52(6), 2069-2078 (2008-04-02)
To gain further insight into the understanding of the antiviral resistance patterns and mechanisms of the integrase strand transfer inhibitor L-870,810, the prototypical naphthyridine analogue, we passaged the human immunodeficiency virus type 1 strain HIV-1(III(B)) in cell culture in the
Motohide Sato et al.
Journal of medicinal chemistry, 49(5), 1506-1508 (2006-03-03)
The viral enzyme integrase is essential for the replication of human immunodeficiency virus type 1 (HIV-1) and represents a remaining target for antiretroviral drugs. Here, we describe the modification of a quinolone antibiotic to produce the novel integrase inhibitor JTK-303
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SML3672-50MG | 04065269082743 |
| SML3672-10MG | 04065269082736 |
